Assay Details
| Gene Symbol: Gm13857 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK133155.1 | - | 594 | 84 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
aldo-keto reductase family 1, member B7 |
Akr1b7 |
AR, Avdp, MVDP |
Anti-sense | Chr. 6 - 34412362 - 34423137 on NCBI Build GRCm38 | |
| Downstream | 2,3-bisphosphoglycerate mutase | Bpgm |
AI323730, AL022789, C86192 |
34.1kb | Anti-sense | Chr. 6 - 34476254 - 34505610 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm13857
Genomic Map