Assay Details
| Gene Symbol: Gm31324 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK133279.1 | - | 1 - 2 | 189 | 97 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 13481 | Gm13481 | OTTMUSG00000012370 | -636.8kb | Anti-sense | Chr. 2 - 48062162 - 48156559 on NCBI Build GRCm38 |
| Downstream | activin receptor IIA | Acvr2a |
ActrIIa, Acvr2, TactrII |
19.5kb | Sense | Chr. 2 - 48814109 - 48903264 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm31324
Genomic Map