Assay Details
| Gene Symbol: Gm33280 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK137601.1 | - | 275 | 78 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | heparan sulfate 6-O-sulfotransferase 1 | Hs6st1 | 6Ost1 | -25.6kb | Sense | Chr. 1 - 36068400 - 36106446 on NCBI Build GRCm38 |
| Downstream | UDP-glucose glycoprotein glucosyltransferase 1 | Uggt1 |
0910001L17Rik, A930007H10Rik, AA589501, AI414429, AI448372, C820010P03Rik, GT, UGT1, Ugcgl1 |
6.9kb | Anti-sense | Chr. 1 - 36140028 - 36244302 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm33280
Genomic Map