Assay Details
| Gene Symbol: Gm16796 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040367.1 | 2 - 3 | 2766 | 91 | ||
| GenBank mRNA | AK138533.1 | - | 2 - 3 | 2767 | 91 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | prefoldin 4 | Pfdn4 |
0610025M10Rik, 1810063B06Rik, C-1, C1, Pfd4 |
Anti-sense | Chr. 2 - 170496428 - 170519070 on NCBI Build GRCm38 | |
| Downstream | RIKEN cDNA 4930470P17 gene | 4930470P17Rik | 75.1kb | Sense | Chr. 2 - 170579418 - 170602017 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm16796
Genomic Map