Assay Details
| Gene Symbol: Gm33432 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK142623.1 | - | 345 | 75 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | patatin-like phospholipase domain containing 5 | Pnpla5 |
4833426H19Rik, GS2L |
-1.7kb | Anti-sense | Chr. 15 - 84112356 - 84127992 on NCBI Build GRCm38 |
| Downstream | patatin-like phospholipase domain containing 3 | Pnpla3 | Adpn | 36.1kb | Sense | Chr. 15 - 84167783 - 84189521 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gm33432
Genomic Map