Assay Details
| Gene Symbol: LOC105247423 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK142879.1 | - | 2 - 3 | 794 | 86 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | cholesterol 25-hydroxylase | Ch25h |
AI462618, m25OH |
-248.0 | Anti-sense | Chr. 19 - 34473783 - 34475135 on NCBI Build GRCm38 |
| Downstream | lysosomal acid lipase A | Lipa |
AA960673, Lal, Lip-1, Lip1 |
10.8kb | Anti-sense | Chr. 19 - 34492316 - 34527474 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC105247423
Genomic Map