Assay Details
Gene Symbol: Gm13269 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK146025.1 | - | 2 - 3 | 366 | 92 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | predicted gene 10848 | Gm10848 | ENSMUSG00000075523 | -632.0 | Sense | Chr. 2 - 14709329 - 14739758 on NCBI Build GRCm38 |
Downstream | NOL1/NOP2/Sun domain family member 6 | Nsun6 |
4933403D21Rik, 4933414E04Rik, NOPD1 |
245.4kb | Sense | Chr. 2 - 14991522 - 15054872 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
