Assay Details
| Gene Symbol: Gm26995 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK146140.1 | - | 314 | 101 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
nitric oxide synthase 1, neuronal |
Nos1 |
2310005C01Rik, N-NOS, NC-NOS, NO, NOS, NOS-I, Nos-1, bNOS, nNOS |
Anti-sense | Chr. 5 - 117780997 - 117958840 on NCBI Build GRCm38 | |
| Downstream | F-box protein 21 | Fbxo21 |
2810425J22Rik, AU016673, mKIAA0875 |
38.1kb | Anti-sense | Chr. 5 - 117976735 - 118010201 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gm26995
Genomic Map