Assay Details
| Gene Symbol: Numb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK164313.1 | - | 1 | 3048 | 71 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
papilin, proteoglycan-like sulfated glycoprotein |
Papln | E030033C16Rik | -27.7kb | Anti-sense | Chr. 12 - 83762992 - 83794875 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 2410016O06 gene | 2410016O06Rik |
MAPJD, NO66 |
122.9kb | Anti-sense | Chr. 12 - 83950608 - 83952953 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Numb
Genomic Map