Assay Details
| Gene Symbol: Gm38575 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK038448.1 | - | 245 | 98 | ||
| AK165297.1 | - | 176 | 98 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | cryptochrome 1 (photolyase-like) | Cry1 |
AU020726, AU021000, Phll1 |
-46.5kb | Sense | Chr. 10 - 85131700 - 85185054 on NCBI Build GRCm38 |
| Downstream | BTB (POZ) domain containing 11 | Btbd11 | 6330404E16Rik | 120.4kb | Anti-sense | Chr. 10 - 85386776 - 85660292 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gm38575
Genomic Map