Assay Details
| Gene Symbol: Ccdc134 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC004684.1 | - | 1 | 1045 | 98 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | meiosis defective 1 | Mei1 |
4932408F18Rik, mei1 |
-8.1kb | Sense | Chr. 15 - 82070059 - 82126817 on NCBI Build GRCm38 |
| Downstream | sterol regulatory element binding factor 2 | Srebf2 |
AI608257, SREBP-2, SREBP2, SREBP2gc, bHLHd2, lop13, nuc |
11.2kb | Sense | Chr. 15 - 82147269 - 82205379 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ccdc134
Genomic Map