Assay Details
| Gene Symbol: Gm27008 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK038087.1 | - | 164 | 70 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 35507 |
Gm35507 | -70.1kb | Sense | Chr. 3 - 106827307 - 106879361 on NCBI Build GRCm38 | |
| Downstream |
potassium voltage-gated channel, shaker-related subfamily, member 2 |
Kcna2 |
Akr6a4, ENSMUSG00000074335, Gm10672, Kca1-2, Kv1.2, Mk-2 |
35.3kb | Anti-sense | Chr. 3 - 107090852 - 107115005 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm27008
Genomic Map