Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | like-glycosyltransferase | Large |
BPFD#36, Gyltl1a, Mbp-1, Mbp1, enr, fg, froggy, mKIAA0609, myd |
Sense | Chr. 8 - 72814599 - 73353577 on NCBI Build GRCm38 | |
| Downstream | microRNA 28b | Mir28b | mmu-mir-28b | 63.8kb | Anti-sense | Chr. 8 - 73016512 - 73016591 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map