Assay Details
| Gene Symbol: LOC102635358 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK038608.1 | - | 2 - 3 | 240 | 87 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
leucine-rich repeat LGI family, member 1 |
Lgi1 | BB130740 | -209.0 | Anti-sense | Chr. 19 - 38264782 - 38308939 on NCBI Build GRCm38 |
| Downstream |
predicted gene, 32960 |
Gm32960 | 36.9kb | Anti-sense | Chr. 19 - 38348636 - 38350079 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

LOC102635358
Genomic Map