Assay Details
| Gene Symbol: Lipt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK039030.1 | - | 210 | 68 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | testis specific 10 | Tsga10 |
4933432N21Rik, AI843383, Gm217, Mtsga10 |
Anti-sense | Chr. 1 - 37728144 - 37866739 on NCBI Build GRCm38 | |
| Downstream |
MIT, microtubule interacting and transport, domain containing 1 |
Mitd1 | 1500032H18Rik | 10.1kb | Anti-sense | Chr. 1 - 37878890 - 37890428 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Lipt1
Genomic Map