Assay Details
| Gene Symbol: Gm13920 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK039323.1 | - | 1 - 2 | 165 | 94 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 39888 |
Gm39888 | -44.8kb | Sense | Chr. 2 - 101982108 - 102012789 on NCBI Build GRCm38 | |
| Downstream | tripartite motif-containing 44 | Trim44 |
Dipb, Mc7 |
238.7kb | Anti-sense | Chr. 2 - 102300119 - 102400900 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm13920
Genomic Map