Assay Details
| Gene Symbol: Gm29927 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK039399.1 | - | 1 - 2 | 79 | 100 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
phosphatidylinositol 3-kinase, regulatory subunit, polypeptide 1 (p85 alpha) |
Pik3r1 |
PI3K, p50alpha, p55alpha, p85alpha |
-795.0kb | Sense | Chr. 13 - 101680761 - 101768217 on NCBI Build GRCm38 |
| Downstream | CD180 antigen | Cd180 |
F630107B15, Ly78, RP105 |
103.3kb | Anti-sense | Chr. 13 - 102693558 - 102706631 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm29927
Genomic Map