Assay Details
| Gene Symbol: Gm16793 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040376.1 | 2 - 3 | 471 | 96 | ||
| GenBank mRNA | AK040224.1 | - | 2 - 3 | 424 | 96 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | exoribonuclease 1 | Eri1 |
3'hexo, 3110010F15Rik, Thex1, eri-1 |
-87.0kb | Sense | Chr. 8 - 35465265 - 35495533 on NCBI Build GRCm38 |
| Downstream | malignant fibrous histiocytoma amplified sequence 1 | Mfhas1 |
2310066G09Rik, D8Ertd91e |
3.2kb | Anti-sense | Chr. 8 - 35587777 - 35679449 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm16793
Genomic Map