Assay Details
| Gene Symbol: Gm16725 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK040746.1 | - | 3 - 4 | 639 | 90 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
microcephaly, primary autosomal recessive 1 |
Mcph1 |
5430437K10Rik, BRIT1, D030046N04Rik, MCT |
Anti-sense | Chr. 8 - 18595136 - 18803189 on NCBI Build GRCm38 | |
| Downstream | angiopoietin 2 | Angpt2 |
Agpt2, Ang-2, Ang2 |
36.7kb | Sense | Chr. 8 - 18690263 - 18741562 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm16725
Genomic Map