Assay Details
| Gene Symbol: Gm15551 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK041310.1 | - | 1 - 2 | 185 | 69 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | arylsulfatase J | Arsj |
9330196J05Rik, D830047F08 |
-227.0kb | Anti-sense | Chr. 3 - 126363250 - 126440379 on NCBI Build GRCm38 |
| Downstream |
ankyrin 2, brain |
Ank2 |
AI835472, AW491075, Ank-2 |
252.6kb | Sense | Chr. 3 - 126921607 - 127499870 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gm15551
Genomic Map