Assay Details
| Gene Symbol: Gm32548 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK042350.1 | - | 2 - 3 | 280 | 77 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | SMAD family member 4 | Smad4 |
AW743858, D18Wsu70e, DPC4, Madh4 |
-11.1kb | Anti-sense | Chr. 18 - 73638998 - 73703741 on NCBI Build GRCm38 |
| Downstream | elaC homolog 1 (E. coli) | Elac1 |
2610018O07Rik, 8430417G19Rik |
17.4kb | Anti-sense | Chr. 18 - 73735038 - 73754758 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm32548
Genomic Map