Assay Details
| Gene Symbol: LOC102642641 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK042451.1 | - | 1 - 2 | 1299 | 186 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
solute carrier organic anion transporter family, member 3a1 |
Slco3a1 |
5830414C08Rik, Anr1, MJAM, OATP-D, R75096, Slc21a11 |
-3.2kb | Anti-sense | Chr. 7 - 74275418 - 74554780 on NCBI Build GRCm38 |
| Downstream |
predicted gene, 30459 |
Gm30459 | 206.1kb | Sense | Chr. 7 - 74765987 - 74771243 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC102642641
Genomic Map