Assay Details
| Gene Symbol: Gm29128 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK139037.1 | - | 1 - 2 | 73 | 78 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 29669 | Gm29669 | Anti-sense | Chr. 1 - 31117532 - 31135201 on NCBI Build GRCm38 | ||
| Downstream | RIKEN cDNA 4931428L18 gene | 4931428L18Rik | 4930563O14Rik | 2.8kb | Anti-sense | Chr. 1 - 31141078 - 31222656 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm29128
Genomic Map