Assay Details
| Gene Symbol: Gm35769 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK143082.1 | - | 6 - 7 | 684 | 105 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | cadherin 18 | Cdh18 |
B230220E17Rik, Cdh14l |
-1.4mb | Anti-sense | Chr. 15 - 22548899 - 23474432 on NCBI Build GRCm38 |
| Downstream | predicted gene 2824 | Gm2824 | 122.2kb | Sense | Chr. 15 - 24978422 - 25027708 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm35769
Genomic Map