Assay Details
| Gene Symbol: Gm26574 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK143393.1 | - | 1 - 2 | 364 | 89 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
protein tyrosine phosphatase, non-receptor type 14 |
Ptpn14 |
C130080N23Rik, OTTMUSG00000022087, PTP36 |
Anti-sense | Chr. 1 - 189728216 - 189876695 on NCBI Build GRCm38 | |
| Downstream | SET and MYND domain containing 2 | Smyd2 |
1110020E07Rik, 4930402C15, KMT3C, Zmynd14 |
43.9kb | Sense | Chr. 1 - 189880492 - 189922379 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm26574
Genomic Map