Assay Details
| Gene Symbol: Gm33474 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK143409.1 | - | 1 - 2 | 624 | 112 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | cell division cycle 7 (S. cerevisiae) | Cdc7 |
AI597260, Cdc7l1, muCdc7 |
-91.4kb | Sense | Chr. 5 - 106964315 - 106984440 on NCBI Build GRCm38 |
| Downstream |
transforming growth factor, beta receptor III |
Tgfbr3 |
1110036H20Rik, AU015626, AW215636, TBRIII |
27.8kb | Anti-sense | Chr. 5 - 107106570 - 107289595 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm33474
Genomic Map