Assay Details
| Gene Symbol: Gm14487 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK146369.1 | - | 2 - 3 | 418 | 81 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | immediate early response 5-like | Ier5l | 2610524G09Rik | -62.2kb | Anti-sense | Chr. 2 - 30472631 - 30474199 on NCBI Build GRCm38 |
| Downstream |
CSA-conditional, T cell activation-dependent protein |
Cstad | 9530039B02Rik | 56.0kb | Sense | Chr. 2 - 30595044 - 30608945 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm14487
Genomic Map