Assay Details
| Gene Symbol: Gm16731 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK164782.1 | - | 161 | 74 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
coronin, actin binding protein 2A |
Coro2a |
9030208C03Rik, AI563590, IR10 |
Anti-sense | Chr. 4 - 46536937 - 46601929 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm16731
Genomic Map