Assay Details
| Gene Symbol: Gm16364 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK045600.1 | - | 1 - 2 | 28 | 78 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | Fyn proto-oncogene | Fyn |
AI448320, AW552119 |
Anti-sense | Chr. 10 - 39369770 - 39565381 on NCBI Build GRCm38 | |
| Downstream |
predicted gene, 40627 |
Gm40627 | 32.4kb | Anti-sense | Chr. 10 - 39590365 - 39599092 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm16364
Genomic Map