Assay Details
| Gene Symbol: Gm17597 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK045688.1 | - | 1 - 2 | 202 | 88 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc finger CCCH type containing 7B | Zc3h7b |
Gm627, Scrg3, Zc2h7b, mKIAA1031 |
-4.5kb | Anti-sense | Chr. 15 - 81744848 - 81796269 on NCBI Build GRCm38 |
| Downstream | thyrotroph embryonic factor | Tef | 2310028D20Rik | 60.0 | Anti-sense | Chr. 15 - 81802673 - 81826863 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gm17597
Genomic Map