Assay Details
| Gene Symbol: Gm30329 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK045708.1 | - | 1 - 2 | 139 | 70 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc finger protein 827 | Zfp827 |
2810449M09Rik, D630040G17Rik, Znf827 |
-32.2kb | Sense | Chr. 8 - 79027983 - 79193768 on NCBI Build GRCm38 |
| Downstream | methylmalonic aciduria (cobalamin deficiency) type A | Mmaa |
2810018E08Rik, AI840684 |
33.4kb | Anti-sense | Chr. 8 - 79263597 - 79294973 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm30329
Genomic Map