Assay Details
| Gene Symbol: Cdc37l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK046873.1 | - | 1 | 2948 | 93 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | adenylate kinase 3 | Ak3 |
AA407498, AI506714, AK-3, Ak3l, Ak3l1, Akl3l |
Anti-sense | Chr. 19 - 28995380 - 29048672 on NCBI Build GRCm38 | |
| Downstream | RIKEN cDNA 1700018L02 gene | 1700018L02Rik | 27.3kb | Sense | Chr. 19 - 29047483 - 29048729 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Cdc37l1
Genomic Map