Assay Details
| Gene Symbol: Zak | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164791.1 | NP_001158263.1 | 12 | 6031 | 90 | |
| GenBank mRNA | AK051330.1 | - | 1 | 246 | 90 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | Rap guanine nucleotide exchange factor (GEF) 4 | Rapgef4 |
1300003D15Rik, 5730402K07Rik, 6330581N18Rik, EPAC 2, Epac-2, Epac2, mKIAA4040 |
-149.1kb | Sense | Chr. 2 - 71981215 - 72257474 on NCBI Build GRCm38 |
| Downstream | cell division cycle associated 7 | Cdca7 |
2310021G01Rik, AU044109, JPO1 |
68.7kb | Sense | Chr. 2 - 72476189 - 72486892 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Zak
Genomic Map