Assay Details
| Gene Symbol: Gm13571 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK052471.1 | - | 3 - 4 | 445 | 94 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | CD302 antigen | Cd302 |
1110055L24Rik, AI159627 |
-71.4kb | Anti-sense | Chr. 2 - 60251993 - 60284484 on NCBI Build GRCm38 |
| Downstream | predicted gene 13580 | Gm13580 | OTTMUSG00000012842 | 50.4kb | Sense | Chr. 2 - 60411525 - 60412574 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm13571
Genomic Map