Assay Details
| Gene Symbol: Gm14261 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK052878.1 | - | 1 - 2 | 104 | 105 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
ATPase, class II, type 9A |
Atp9a | IIa | -24.0kb | Anti-sense | Chr. 2 - 168634438 - 168742136 on NCBI Build GRCm38 |
| Downstream | zinc finger protein 64 | Zfp64 | 157.3kb | Anti-sense | Chr. 2 - 168925361 - 168955587 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gm14261
Genomic Map