Assay Details
| Gene Symbol: Gm29601 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK076723.1 | - | 1 - 2 | 79 | 95 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 1700063A18 gene | 1700063A18Rik | -521.3kb | Sense | Chr. 1 - 95990659 - 96021758 on NCBI Build GRCm38 | |
| Downstream | RIKEN cDNA 4930533P14 gene | 4930533P14Rik | 55.4kb | Sense | Chr. 1 - 96612057 - 96662573 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm29601
Genomic Map