Assay Details
| Gene Symbol: Gm34299 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK077353.1 | - | 4 - 5 | 1102 | 69 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 3085 | Mir3085 |
mir-3085, mmu-mir-3085 |
-132.8kb | Anti-sense | Chr. 19 - 42280082 - 42280170 on NCBI Build GRCm38 |
| Downstream | R3H domain and coiled-coil containing 1 like | R3hcc1l |
1700036B12Rik, AI450607, C10orf28, D19Ertd386e, GIDRP86, Gidrp88 |
104.1kb | Sense | Chr. 19 - 42518312 - 42592256 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm34299
Genomic Map