Assay Details
| Gene Symbol: Gm33867 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK134227.1 | - | 111 | 79 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 6260 | Gm6260 | EG621809 | -31.2kb | Anti-sense | Chr. 3 - 143670892 - 143712143 on NCBI Build GRCm38 |
| Downstream |
predicted gene, 34078 |
Gm34078 | 331.2kb | Anti-sense | Chr. 3 - 144132352 - 144136012 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm33867
Genomic Map