Assay Details
| Gene Symbol: Gm26908 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK134715.1 | - | 657 | 77 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | sorbin and SH3 domain containing 3 | Sorbs3 |
SCAM-1, SH3P3, Sh3d4, vinexin-g |
Anti-sense | Chr. 14 - 70180468 - 70212457 on NCBI Build GRCm38 | |
| Downstream |
protein phosphatase 3, catalytic subunit, gamma isoform |
Ppp3cc |
Calnc, PP2BA gamma |
9.0kb | Anti-sense | Chr. 14 - 70217864 - 70289508 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm26908
Genomic Map