Assay Details
| Gene Symbol: Gm34068 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK139592.1 | - | 2 - 3 | 303 | 65 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | hedgehog interacting protein-like 2 | Hhipl2 |
4930507C10Rik, AW208802, Hhip3 |
-345.1kb | Sense | Chr. 1 - 183413929 - 183437053 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 1700056E22 gene | 1700056E22Rik | 223.3kb | Anti-sense | Chr. 1 - 184033032 - 184033998 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm34068
Genomic Map