Assay Details
| Gene Symbol: Gm16036 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK139684.1 | - | 273 | 70 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
caspase recruitment domain family, member 11 |
Card11 |
0610008L17Rik, 2410011D02Rik, BIMP3, CARMA1 |
-595.8kb | Sense | Chr. 5 - 140872999 - 141000596 on NCBI Build GRCm38 |
| Downstream | forkhead box K1 | Foxk1 |
A630048H08Rik, AI463295, ENSMUSG00000075577, Gm10868, Mnf |
794.9kb | Anti-sense | Chr. 5 - 142401497 - 142462015 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm16036
Genomic Map