Assay Details
Gene Symbol: Gm30844 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK139796.1 | - | 1 - 2 | 43 | 74 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | RIKEN cDNA 4732490B19 gene | 4732490B19Rik | -30.4kb | Anti-sense | Chr. 11 - 113203315 - 113209669 on NCBI Build GRCm38 | |
Downstream |
solute carrier family 39 (metal ion transporter), member 11 |
Slc39a11 | 1810074D23Rik | 4.4kb | Anti-sense | Chr. 11 - 113244851 - 113650752 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
