Assay Details
| Gene Symbol: Gm30844 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK139796.1 | - | 2 - 3 | 164 | 100 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 4732490B19 gene | 4732490B19Rik | -30.7kb | Anti-sense | Chr. 11 - 113203315 - 113209669 on NCBI Build GRCm38 | |
| Downstream | somatostatin receptor 2 | Sstr2 |
SSTR-2, Smstr-2, Smstr2, sst2 |
319.1kb | Sense | Chr. 11 - 113619481 - 113625895 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm30844
Genomic Map