Assay Details
| Gene Symbol: Gm12126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK139851.1 | - | 1 - 2 | 373 | 80 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | teneurin transmembrane protein 2 | Tenm2 |
Odz2, Odz3, Ten-2, Ten-m2 |
Anti-sense | Chr. 11 - 36006656 - 37236808 on NCBI Build GRCm38 | |
| Downstream | predicted gene 12128 | Gm12128 | OTTMUSG00000005422 | 1.6mb | Sense | Chr. 11 - 37658989 - 37673442 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gm12126
Genomic Map