Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1200016352.mm.1 |
NR_045751.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200016352.mm.1 |
NR_045751.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200016352.mm.1 |
TC1200002351.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200016356.mm.1 |
NR_045751.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200016356.mm.1 |
NR_045751.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200016356.mm.1 |
NR_045752.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200016356.mm.1 |
NR_045752.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200016356.mm.1 |
TC1200002351.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200016358.mm.1 |
NR_045751.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200016358.mm.1 |
NR_045751.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200016358.mm.1 |
NR_045752.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200016358.mm.1 |
NR_045752.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200016358.mm.1 |
NR_045753.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200016358.mm.1 |
NR_045753.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200016358.mm.1 |
TC1200002351.mm.1 |
MTA TC |