Assay Details
| Gene Symbol: Gm13293 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040369.1 | 2 - 3 | 499 | 81 | ||
| GenBank mRNA | AK140161.1 | - | 2 - 3 | 499 | 81 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
protein kinase C, theta |
Prkcq |
A130035A12Rik, AW494342, PKC-0, PKC-theta, PKCtheta, Pkcq |
-40.2kb | Sense | Chr. 2 - 11172382 - 11301226 on NCBI Build GRCm38 |
| Downstream | 6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 3 | Pfkfb3 |
E330010H22Rik, iPFK-2, uPFK-2 |
128.8kb | Anti-sense | Chr. 2 - 11471430 - 11554071 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm13293
Genomic Map