Assay Details
| Gene Symbol: Gm26534 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK140192.1 | - | 236 | 54 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | transmembrane protein 11 | Tmem11 |
5730466P16Rik, AA409091 |
-80.0 | Anti-sense | Chr. 11 - 60864452 - 60879038 on NCBI Build GRCm38 |
| Downstream | N-acetyltransferase domain containing 1 | Natd1 |
AI256713, F3-2, Gm16515, Gtlf3b |
16.1kb | Anti-sense | Chr. 11 - 60902246 - 60913792 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm26534
Genomic Map