Assay Details
| Gene Symbol: Gm15083 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK140501.1 | - | 1 - 2 | 76 | 76 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | SEC13 homolog (S. cerevisiae) | Sec13 |
1110003H02Rik, Sec13l1, Sec13r |
-264.1kb | Anti-sense | Chr. 6 - 113728052 - 113740681 on NCBI Build GRCm38 |
| Downstream |
solute carrier family 6 (neurotransmitter transporter, GABA), member 11 |
Slc6a11 |
D930045G19Rik, E130202I16Rik, GAT4, Gabt4, Gat3 |
80.8kb | Sense | Chr. 6 - 114131241 - 114249886 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm15083
Genomic Map