Assay Details
| Gene Symbol: Gm19710 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045749.1 | 3 - 4 | 423 | 89 | ||
| GenBank mRNA | AK140585.1 | - | 3 - 4 | 425 | 89 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | HCLS1 associated X-1 | Hax1 |
HAX-1, HSP1BP-1, Hs1bp1, SIG-111, Silg111, mHAX-1s |
-1.5kb | Anti-sense | Chr. 3 - 89995446 - 89998716 on NCBI Build GRCm38 |
| Downstream | microRNA 7669 | Mir7669 |
Gm27996, mmu-mir-7669 |
9.8kb | Anti-sense | Chr. 3 - 90011153 - 90011210 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm19710
Genomic Map