Assay Details
| Gene Symbol: Gm10390 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK143646.1 | - | 1 - 2 | 172 | 87 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 38554 |
Gm38554 | -168.1kb | Anti-sense | Chr. 5 - 119959177 - 119963380 on NCBI Build GRCm38 | |
| Downstream | LIM homeobox protein 5 | Lhx5 | Lim2 | 292.6kb | Anti-sense | Chr. 5 - 120431886 - 120441457 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm10390
Genomic Map